Anti-CDNF Rabbit Polyclonal Antibody

Supplier: Sino Biological
50569-RP01-400 50569-RP01-200 50569-RP01-100
103658-398EA 562.97 USD
103658-398 103658-396 103658-394
Anti-CDNF Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CDNF (rM CDNF; Catalog#50569-M08H; NP_808315.1; Met 1-Leu 187). Total IgG was purified by Protein A affinity chromatography
.

Type: Primary
Antigen: CDNF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR