Anti-GPA33 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50559-RP02-100 50559-RP02-50 50559-RP02-200
103660-682EA 270.87 USD
103660-682 103595-202 103660-684
Anti-GPA33 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse GPA33 (rM GPA33; Catalog#50559-M08H; NP_067623.1; Met 1-Ile 235). GPA33 specific IgG was purified by Mouse GPA33 affinity chromatography.

Type: Primary
Antigen: A33
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR