Anti-GPA33 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50559-RP01-100 50559-RP01-200 50559-RP01-400
103658-380EA 270.2 USD
103658-380 103658-384 103658-382
Anti-GPA33 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse GPA33 (rM GPA33; Catalog#50559-M08H; NP_067623.1; Met 1-Ile 235). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: A33
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR