Anti-LYPD3 Rabbit Polyclonal Antibody

50546-T16-50 50546-T16-200 50546-T16-100
103589-172EA 106.62 USD
103589-172 103594-646 103594-644
Anti-LYPD3 Rabbit Polyclonal Antibody
Anti-LYPD3 Antibody

Type: Primary
Antigen: LYPD3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR