Anti-SPARCL1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50544-RP01-100 50544-RP01-200 50544-RP01-400
103659-378EA 106.62 USD
103659-378 103659-376 103659-380
Anti-SPARCL1 Rabbit Polyclonal Antibody
Antibodies
Anti-SPARCL1 Antibody

Type: Primary
Antigen: SPARCL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR