Anti-ACHE Rabbit Polyclonal Antibody

Supplier: Sino Biological

50543-RP01-200 50543-RP01-100 50543-RP01-400
103659-366EA 160.46 USD
103659-366 103659-368 103659-370
Anti-ACHE Rabbit Polyclonal Antibody
Antibodies
Anti-Acetylcholinesterase Anti

Type: Primary
Antigen: ACHE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR