Anti-DDR2 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50541-RP01-200 50541-RP01-100 50541-RP01-400
103660-680EA 160.46 USD
103660-680 103660-678 103658-330
Anti-DDR2 Rabbit Polyclonal Antibody
Antibodies
Anti-DDR2 Antibody

Type: Primary
Antigen: DDR2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR