Anti-Factor D Rabbit Polyclonal Antibody

Supplier: Sino Biological

50539-T16-200 50539-T16-50 50539-T16-100
103595-176EA 214.31 USD
103595-176 103589-168 103595-174
Anti-Factor D Rabbit Polyclonal Antibody
Antibodies
Anti-Factor D Antibody

Type: Primary
Antigen: Factor D
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR