Anti-CFD Rabbit Monoclonal Antibody [clone:121]

Supplier: Sino Biological
50539-R121-100 50539-R121-50
103595-172EA 150.18 USD
103595-172 103658-324
Anti-CFD Rabbit Monoclonal Antibody [clone:121]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Adipsin / Complement Factor D (Catalog#50539-M08H; P03953-2; Met1-Ser258).

Type: Primary
Antigen: Factor D
Clonality: Monoclonal
Clone: 121
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR