Anti-CFD Rabbit Monoclonal Antibody [clone:104]

Supplier: Sino Biological
50539-R104-50 50539-R104-100
103658-320EA 271.4 USD
103658-320 103595-168
Anti-CFD Rabbit Monoclonal Antibody [clone:104]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Adipsin / Complement Factor D / CFD (rM Adipsin / Complement Factor D / CFD; Catalog#50539-M08H; P03953-2; Met1-Ser258).

Type: Primary
Antigen: Factor D
Clonality: Monoclonal
Clone: 104
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR