Anti-CD2 Rabbit Monoclonal Antibody [clone:029]

Supplier: Sino Biological
50537-R029-100 50537-R029-50
103595-164EA 150.18 USD
103595-164 103658-308
Anti-CD2 Rabbit Monoclonal Antibody [clone:029]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD2 (rM CD2; Catalog#50537-M08H; NP_038514.1; Met 1-Ser 203).

Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: 029
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR