Anti-CRABP2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50535-RP01-100 50535-RP01-200 50535-RP01-400
103658-300EA 351.49 USD
103658-300 103658-298 103658-302
Anti-CRABP2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CRABP2 (Catalog#50535-M07E; P22935; Pro2-Glu138). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CRABP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR