Anti-CSRP1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50532-T52-100 50532-T52-200 50532-T52-50
103589-166EA 142.96 USD
103589-166 103595-158 103595-156
Anti-CSRP1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CSRP1 (Catalog#50532-M08E; P97315; Met1-Glu193). CSRP1 specific IgG was purified by Mouse CSRP1 affinity chromatography.

Type: Primary
Antigen: CSRP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR