Anti-c-Kit Rabbit Polyclonal Antibody

Supplier: Sino Biological

50530-RP02-50 50530-RP02-100 50530-RP02-200
103595-144EA 106.62 USD
103595-144 103658-286 103658-288
Anti-c-Kit Rabbit Polyclonal Antibody
Antibodies
Anti-c-Kit Antibody

Type: Primary
Antigen: c-Kit
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR