Anti-METAP2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50528-RP01-200 50528-RP01-400 50528-RP01-100
103658-268EA 351.49 USD
103658-268 103658-270 103658-272
Anti-METAP2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse MAP2 (Catalog#50528-M08B; O08663; Ala2-Tyr478). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: MAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR