Anti-METAP2 Rabbit Monoclonal Antibody [clone:010]

Supplier: Sino Biological
50528-R010-100 50528-R010-50
103658-266EA 271.4 USD
103658-266 103595-138
Anti-METAP2 Rabbit Monoclonal Antibody [clone:010]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse MAP2 / METAP2 (rM MAP2 / METAP2; Catalog#50528-M08B; O08663; Ala2-Tyr478).

Type: Primary
Antigen: MAP2
Clonality: Monoclonal
Clone: 010
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR