Anti-MAP2 Rabbit Monoclonal Antibody [clone: 001]

Supplier: Sino Biological

50528-R001-100 50528-R001-50
103658-264EA 214.31 USD
103658-264 103595-136
Anti-MAP2 Rabbit Monoclonal Antibody [clone: 001]
Antibodies
Anti-methionyl aminopeptidase

Type: Primary
Antigen: MAP2
Clonality: Monoclonal
Clone: Clone: 001
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR