Anti-CD99L2 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50526-RP02-100 50526-RP02-50 50526-RP02-200
103658-260EA 160.46 USD
103658-260 103595-134 103658-262
Anti-CD99L2 Rabbit Polyclonal Antibody
Antibodies
Anti-CD99L2 Antibody

Type: Primary
Antigen: CD99L2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR