Anti-CD99L2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50526-RP01-400 50526-RP01-200 50526-RP01-100
103658-256EA 351.49 USD
103658-256 103658-254 103658-258
Anti-CD99L2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CD99L2 (rM CD99L2; Catalog#50526-M08H; NP_612182.1; Met 1-Ala 164). Total IgG was purified by Protein A affinity chromatography
.

Type: Primary
Antigen: CD99L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR