Anti-DLL1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50522-RP01-100 50522-RP01-200 50522-RP01-400
103658-244EA 106.62 USD
103658-244 103658-246 103658-248
Anti-DLL1 Rabbit Polyclonal Antibody
Antibodies
Anti-DLL1 Antibody

Type: Primary
Antigen: DLL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR