Anti-DLL1 Rabbit Monoclonal Antibody [clone:036]

Supplier: Sino Biological
50522-R036-100 50522-R036-50
103658-242EA 271.4 USD
103658-242 103595-126
Anti-DLL1 Rabbit Monoclonal Antibody [clone:036]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse DLL1 (rM DLL1; Catalog#50522-M08H; NP_031891.2; Met 1-Gln 516).

Type: Primary
Antigen: DLL1
Clonality: Monoclonal
Clone: 036
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR