Anti-DLL1 Rabbit Monoclonal Antibody [clone: 019]

Supplier: Sino Biological

50522-R019-50 50522-R019-100
103595-124EA 128.15 USD
103595-124 103660-672
Anti-DLL1 Rabbit Monoclonal Antibody [clone: 019]
Antibodies
Anti-DLL1 Antibody

Type: Primary
Antigen: DLL1
Clonality: Monoclonal
Clone: Clone: 019
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR