Anti-CES2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50515-RP02-200 50515-RP02-100 50515-RP02-50
103660-670EA 352.03 USD
103660-670 103660-668 103595-122
Anti-CES2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CES2 / Carboxylesterase 2 (rM CES2 / Carboxylesterase 2; Catalog#50543-M08H; NP_663578.1; Met1-Lys557). CES2 / Carboxylesterase 2 specific IgG was purified by Mouse Acetylcholinesterase / ACHE affinity chromatography.

Type: Primary
Antigen: CES2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR