Anti-LRIG1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50511-T16-50 50511-T16-200 50511-T16-100
103589-160EA 106.62 USD
103589-160 103595-120 103595-118
Anti-LRIG1 Rabbit Polyclonal Antibody
Antibodies
Anti-LRIG1 Antibody

Type: Primary
Antigen: LRIG1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR