Anti-LY9 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50504-RP01-200 50504-RP01-100 50504-RP01-400
103658-232EA 562.97 USD
103658-232 103658-228 103658-230
Anti-LY9 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse LY9 / CD229 / SLAMF3 (rM LY9 / CD229 / SLAMF3; Catalog#50504-M08H; NP_032560.2; Met 1-Arg 453). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CD229
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR