Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD229 Rabbit Monoclonal Antibody [clone: 102]

Supplier: Sino Biological

50504-R102-100 50504-R102-50
103658-226EA 214.31 USD InStock
103658-226 103595-114
Anti-CD229 Rabbit Monoclonal Antibody [clone: 102]
Antibodies
Anti-CD229 Antibody

Type: Primary
Antigen: CD229
Clonality: Monoclonal
Clone: Clone: 102
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty