Anti-CD229 Rabbit Monoclonal Antibody [clone: 017]

Supplier: Sino Biological

50504-R017-100 50504-R017-50
103658-224EA 214.31 USD
103658-224 103595-112
Anti-CD229 Rabbit Monoclonal Antibody [clone: 017]
Antibodies
Anti-CD229 Antibody

Type: Primary
Antigen: CD229
Clonality: Monoclonal
Clone: Clone: 017
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR