Anti-LY9 Rabbit Monoclonal Antibody [clone:002] PerCP (Peridinin-Chlorophyll Protein Complex)

Supplier: Sino Biological
50504-R002-C-25 50504-R002-C-100
103630-088EA 90.9 USD
103630-088 103632-268
Anti-LY9 Rabbit Monoclonal Antibody [clone:002] PerCP (Peridinin-Chlorophyll Protein Complex)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Ly9 / CD229 / SLAMF3 (rM Ly9 / CD229 / SLAMF3; Catalog#50504-M08H; NP_032560.2; Met1-Arg453) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.

Type: Primary
Antigen: CD229 (PerCP)
Clonality: Monoclonal
Clone: Clone: 002
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR