Anti-LY9 Rabbit Monoclonal Antibody [clone:002] APC (Allophycocyanin)

Supplier: Sino Biological
50504-R002-A-100 50504-R002-A-25
103630-086EA 90.9 USD
103630-086 103632-266
Anti-LY9 Rabbit Monoclonal Antibody [clone:002] APC (Allophycocyanin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Ly9 / CD229 / SLAMF3 (rM Ly9 / CD229 / SLAMF3; Catalog#50504-M08H; NP_032560.2; Met1-Arg453) and conjugated with APC under optimum conditions, the unreacted APC was removed.

Type: Primary
Antigen: CD229
Clonality: Monoclonal
Clone: Clone: 002
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR