Anti-CD229 Rabbit Monoclonal Antibody [clone: 002]

Supplier: Sino Biological

50504-R002-100 50504-R002-50
103658-222EA 214.31 USD
103658-222 103595-110
Anti-CD229 Rabbit Monoclonal Antibody [clone: 002]
Antibodies
Anti-CD229 Antibody

Type: Primary
Antigen: CD229
Clonality: Monoclonal
Clone: Clone: 002
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR