Anti-CTLA4 Rabbit Monoclonal Antibody [clone:037]

Supplier: Sino Biological
50503-R037-50 50503-R037-100
103658-216EA 271.4 USD
103658-216 103595-102
Anti-CTLA4 Rabbit Monoclonal Antibody [clone:037]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CTLA-4 / CD152 (rM CTLA-4 / CD152; Catalog#50503-M08H; NP_033973.2; Met1-Phe162).

Type: Primary
Antigen: CTLA-4
Clonality: Monoclonal
Clone: 037
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR