Anti-OSMR Rabbit Polyclonal Antibody

Supplier: Sino Biological
50500-T16-100 50500-T16-200 50500-T16-50
103595-098EA 270.87 USD
103595-098 103589-156 103595-100
Anti-OSMR Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse OSMR / IL-31RB (Catalog#50500-M08H; O70458-1; Met1-Leu738). OSMR / IL-31RB specific IgG was purified by Mouse OSMR / IL-31RB affinity chromatography.

Type: Primary
Antigen: OSMR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR