Anti-OSMR Rabbit Polyclonal Antibody

Supplier: Sino Biological
50500-RP01-400 50500-RP01-200 50500-RP01-100
103658-212EA 157.27 USD
103658-212 103658-210 103658-214
Anti-OSMR Rabbit Polyclonal Antibody
Antibodies
Anti-OSMR Antibody

Type: Primary
Antigen: OSMR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR