Anti-OSMR Rabbit Monoclonal Antibody [clone: 202]

Supplier: Sino Biological

50500-R202-100 50500-R202-50
103658-208EA 214.31 USD
103658-208 103595-096
Anti-OSMR Rabbit Monoclonal Antibody [clone: 202]
Antibodies
Anti-OSMR Antibody

Type: Primary
Antigen: OSMR
Clonality: Monoclonal
Clone: Clone: 202
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR