Anti-TNFRSF1A Rabbit Polyclonal Antibody

Supplier: Sino Biological

50496-RP02-100 50496-RP02-200 50496-RP02-50
103658-204EA 160.46 USD
103658-204 103658-206 103595-094
Anti-TNFRSF1A Rabbit Polyclonal Antibody
Antibodies
Anti-TNFRSF1A Antibody

Type: Primary
Antigen: TNFRSF1A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR