Anti-TNFRSF1A Rabbit Polyclonal Antibody

Supplier: Sino Biological
50496-RP01-400 50496-RP01-200 50496-RP01-100
103658-200EA 270.2 USD
103658-200 103658-198 103658-202
Anti-TNFRSF1A Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse TNFR1 / CD120a / TNFRSF1A (Catalog#50496-M08H; NP_035739.2; Met1-Ala212). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: TNFRSF1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR