Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFRSF1A Rabbit Monoclonal Antibody [clone: 104]

Supplier: Sino Biological

50496-R104-50 50496-R104-1 50496-R104-100
103595-092EA 128.15 USD InStock
103595-092 103589-154 103659-356
Anti-TNFRSF1A Rabbit Monoclonal Antibody [clone: 104]
Antibodies
Anti-TNFRSF1A Antibody

Type: Primary
Antigen: TNFRSF1A
Clonality: Monoclonal
Clone: Clone: 104
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty