Anti-SPARC Rabbit Monoclonal Antibody [clone:001]

Supplier: Sino Biological
50494-R001-100 50494-R001-50
103659-342EA 271.4 USD
103659-342 103595-082
Anti-SPARC Rabbit Monoclonal Antibody [clone:001]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Osteonectin / SPARC (rM Osteonectin / SPARC; Catalog#50494-M08H; NP_033268.1; Met1-Ile302).

Type: Primary
Antigen: SPARC
Clonality: Monoclonal
Clone: 001
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR