Anti-MMP8 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50493-RP01-400 50493-RP01-200 50493-RP01-100
103658-192EA 270.2 USD
103658-192 103658-196 103658-194
Anti-MMP8 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse MMP-8 / MMP8 (Catalog#50493-M08S; NP_032637.3; Met1-Ser465). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: MMP-8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR