Anti-CD180 Rabbit Monoclonal Antibody [clone:105]

Supplier: Sino Biological
50491-R105-100 50491-R105-50
103595-072EA 150.18 USD
103595-072 103658-190
Anti-CD180 Rabbit Monoclonal Antibody [clone:105]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD180 / RP105 (rM CD180 / RP105; Catalog#50491-M08H; NP_032559.2; Met1-Ser626).

Type: Primary
Antigen: RP105
Clonality: Monoclonal
Clone: 105
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR