Anti-CD209B Rabbit Polyclonal Antibody

Supplier: Sino Biological
50486-RP02-50 50486-RP02-200 50486-RP02-100
103595-064EA 142.96 USD
103595-064 103658-170 103658-172
Anti-CD209B Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse SIGNR1 / CD209b (rM SIGNR1 / CD209b; Catalog#50486-M07H; NP_081248.2; Gln 74-Gly 325). SIGNR1 / CD209b specific IgG was purified by Mouse SIGNR1 / CD209b affinity chromatography.

Type: Primary
Antigen: SIGNR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR