Anti-SIGNR1 Rabbit Monoclonal Antibody [clone: 001]

Supplier: Sino Biological

50486-R001-1 50486-R001-100 50486-R001-50
103588-710EA 1281.54 USD
103588-710 103658-158 103595-058
Anti-SIGNR1 Rabbit Monoclonal Antibody [clone: 001]
Antibodies
Anti-SIGNR1 Antibody

Type: Primary
Antigen: SIGNR1
Clonality: Monoclonal
Clone: Clone: 001
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR