Anti-CLU Rabbit Polyclonal Antibody

Supplier: Sino Biological
50485-T16-200 50485-T16-100 50485-T16-50
103588-708EA 142.96 USD
103588-708 103595-054 103595-056
Anti-CLU Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse Clusterin / Apolipoprotein J / Apo-J (rM Clusterin / Apolipoprotein J / Apo-J; Catalog#50485-M08H; NP_038520.2; Met1-Glu448). Clusterin / Apolipoprotein J / Apo-J specific IgG was purified by Mouse Clusterin / Apolipoprotein J / Apo-J affinity chromatography.

Type: Primary
Antigen: Clusterin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR