Anti-S100A4 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50484-T48-100 50484-T48-200 50484-T48-50
103595-048EA 160.46 USD
103595-048 103595-050 103588-706
Anti-S100A4 Rabbit Polyclonal Antibody
Antibodies
Anti-S100A4 Antibody

Type: Primary
Antigen: S100A4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR