Anti-EGF Rabbit Polyclonal Antibody

Supplier: Sino Biological
50482-T16-50 50482-T16-100 50482-T16-200
103595-040EA 270.87 USD
103595-040 103595-042 103589-142
Anti-EGF Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse EGF / Epidermal Growth Factor (Catalog#50482-M08H1; AAH60741.1; Met1-Arg1029). EGF / Epidermal Growth Factor specific IgG was purified by Mouse EGF / Epidermal Growth Factor affinity chromatography.

Type: Primary
Antigen: EGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR