Anti-KNG1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50474-T16-200 50474-T16-100 50474-T16-50
103595-028EA 214.31 USD
103595-028 103595-026 103589-138
Anti-KNG1 Rabbit Polyclonal Antibody
Antibodies
Anti-Kininogen 1 Antibody

Type: Primary
Antigen: KNG1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR