Anti-EGFL7 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50472-RP01-400 50472-RP01-100 50472-RP01-200
103658-128EA 270.2 USD
103658-128 103658-126 103658-130
Anti-EGFL7 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse EGFL7 / VE-statin (Catalog#50472-M07B; Q9QXT5-1; Glu22-Leu275). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: EGFL7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR