Anti-IFNAR1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50469-RP02-50 50469-RP02-100 50469-RP02-200
103658-124EA 352.03 USD
103658-124 103658-122 103595-020
Anti-IFNAR1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse IFNAR-1 / CRF2-1 (rM IFNAR-1; Catalog#50469-M08H; NP_034638.2; Met 1-Thr 429). IFNAR-1 specific IgG was purified by mouse IFNAR-1 affinity chromatography
.

Type: Primary
Antigen: IFNAR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR