Anti-IFNAR1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50469-RP01-400 50469-RP01-100 50469-RP01-200
103659-324EA 99 USD
103659-324 103658-118 103658-120
Anti-IFNAR1 Rabbit Polyclonal Antibody
Antibodies
Anti-IFNAR1 Antibody

Type: Primary
Antigen: IFNAR1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR