Anti-IFNAR1 Rabbit Monoclonal Antibody [clone:114]

Supplier: Sino Biological
50469-R114-100 50469-R114-50
103659-320EA 271.4 USD
103659-320 103595-016
Anti-IFNAR1 Rabbit Monoclonal Antibody [clone:114]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IFNAR1 (rM IFNAR1; Catalog#50469-M08H; NP_034638.2; Met 1-Thr 429).

Type: Primary
Antigen: IFNAR1
Clonality: Monoclonal
Clone: 114
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR